accessibility menu, dialog, popup

UserName

H3COBrNH2

CAS RN 6358-77-6

IUPAC Name: 5-bromo-2-methoxyaniline
Synonyms: 5-bromo-2-methoxyaniline
Molecular Weight (g/mol): 202.05
Molecular Formula: C7H8BrNO
InChi Key: OPGNSNDTPPIYPG-UHFFFAOYSA-N
SMILES: COC1=C(N)C=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results