accessibility menu, dialog, popup

UserName

ONOClO

CAS RN 6361-22-4

IUPAC Name: 2-chloro-6-nitrobenzaldehyde
Synonyms: 2-chloro-6-nitrobenzaldehyde
Molecular Weight (g/mol): 185.56
Molecular Formula: C7H4ClNO3
InChi Key: RZDOUWDCYULHJX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(Cl)=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results