accessibility menu, dialog, popup

UserName

H3COOON

CAS RN 6362-63-6

IUPAC Name: ethyl 3-cyano-2-oxo-3-phenylpropanoate
Synonyms: ethyl 3-cyano-2-oxo-3-phenylpropanoate
Molecular Weight (g/mol): 217.22
Molecular Formula: C12H11NO3
InChi Key: YWNRATDDUSMBPR-UHFFFAOYNA-N
SMILES: CCOC(=O)C(=O)C(C#N)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results