accessibility menu, dialog, popup

UserName

HOO

CAS RN 637-44-5

IUPAC Name: 3-phenylprop-2-ynoic acid
Synonyms: phenylpropiolic acid phenylpropioplate phenylpropioplic acid
Molecular Weight (g/mol): 146.15
Molecular Formula: C9H6O2
InChi Key: XNERWVPQCYSMLC-UHFFFAOYSA-N
SMILES: OC(=O)C#CC1=CC=CC=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results