missing translation for 'onlineSavingsMsg'
Learn More

Phenylpropiolic acid, 98+%

Product Code. 11452158
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452158 50 g 50g
11442158 10 g 10g
11474030 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452158 Supplier Thermo Scientific Alfa Aesar Supplier No. A15163.18
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 637-44-5 | C9H6O2 | 146.15 g/mol

Phenylpropiolic acid is used as a Pharmaceutical Intermediates, laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylpropiolic acid is used as a Pharmaceutical Intermediates, laboratory chemicals.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-44-5
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.15
MDL Number MFCD00004361
InChI Key XNERWVPQCYSMLC-UHFFFAOYSA-N
Synonym phenylpropiolic acid, 3-phenylpropiolic acid, phenylpropynoic acid, 3-phenylpropynoic acid, 2-propynoic acid, 3-phenyl, phenylacetylenecarboxylic acid, 3-phenyl-2-propynoic acid, phenylacetylene monocarboxylic acid, propiolic acid, 3-phenyl, phenyl propiolic acid
PubChem CID 69475
IUPAC Name 3-phenylprop-2-ynoic acid
SMILES OC(=O)C#CC1=CC=CC=C1

Specifications

Melting Point 135°C to 138°C
Quantity 50 g
Beilstein 742587
Solubility Information Soluble in water.
Formula Weight 146.15
Percent Purity ≥98%
Chemical Name or Material Phenylpropiolic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.