accessibility menu, dialog, popup

UserName

H3COClCH3H2N

CAS RN 6376-14-3

IUPAC Name: 4-chloro-2-methoxy-5-methylaniline
Synonyms: 4-chloro-2-methoxy-5-methylaniline
Molecular Weight (g/mol): 171.62
Molecular Formula: C8H10ClNO
InChi Key: XBAPOWUMJRIKAV-UHFFFAOYSA-N
SMILES: COC1=CC(Cl)=C(C)C=C1N
Grade 
  • (2)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results