accessibility menu, dialog, popup

UserName

SNHSNHSHN

CAS RN 638-16-4

IUPAC Name: 1,3,5-triazinane-2,4,6-trithione
Synonyms: 1,3,5-triazinane-2,4,6-trithione
Molecular Weight (g/mol): 177.26
Molecular Formula: C3H3N3S3
InChi Key: WZRRRFSJFQTGGB-UHFFFAOYSA-N
SMILES: S=C1NC(=S)NC(=S)N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)