missing translation for 'onlineSavingsMsg'
Learn More

Trithiocyanuric Acid, TRC

Product Code. 30449798
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449798 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449798 Supplier Toronto Research Chemicals Supplier No. TRCT889305500MG
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trithiocyanuric Acid
CAS 638-16-4
Molecular Formula C3H3N3S3
Quantity 500 mg
Synonym Trithiocyanuric acid','Cyanuric acid','trithio- (6CI), s-Triazine-2,4,6(1H,3H,5H)-trithione (7CI), s-Triazine-2,4,6-trithiol (8CI), 1,3,5-Triazinane-2,4,6-trithione, 1,3,5-Triazine-2,4,6-trithiol, 1,3,5-Triazine-2,4,6-trithione, 1,3,5-Trimercaptotriazine, 2,4,6-Triazinetrithiol, 2,4,6-Trimercapto-1,3,5-triazine, 2,4,6-Trimercapto-s-triazine, 2,4,6-Trimercaptotriazine, 2,4,6-Trithio-5-azauracil, 2,4,6-Trithiol-1,3,5-triazine, Actor TSH, Curekind TCY, Nocceler TCA, NSC 62071, NSC 65480, OF 100, Rhenogran Triazine TM 70AEMD, Rhenogran Triazine TM 70ECO, s-Triazinetrithiol, Taicros TMT, Tatcross TMT, TCY, TCY-MBA, Thiocyanuric acid, TM 70ECO, TMT, Trimercapto-s-triazine, Trimercaptocyanuric acid, Trimercaptotriazine, Trithiocyanuric acid, TSH, Zeonet A, Zisnet F, Zisnet F-ET, Zisnet FPT
InChI Formula InChI=1S/C3H3N3S3/c7-1-4-2(8)6-3(9)5-1/h(H3,4,5,6,7,8,9)
SMILES S=C1NC(=S)NC(=S)N1
IUPAC Name 1,3,5-triazinane-2,4,6-trithione
Molecular Weight (g/mol) 177.27
Formula Weight 176.9489
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.