accessibility menu, dialog, popup

UserName

HClSnH

CAS RN 639-58-7

IUPAC Name: triphenyltin chloride
Synonyms: triphenyltin chloride
Molecular Weight (g/mol): 387.49
Molecular Formula: C18H17ClSn
InChi Key: ZDYYCRIXXMWWQC-UHFFFAOYSA-N
SMILES: Cl.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)