CAS RN 63968-84-3
IUPAC Name:
N'-hydroxy-2-(trifluoromethoxy)benzene-1-carboximidamide
Synonyms:
N'-hydroxy-2-(trifluoromethoxy)benzenecarboximidamide
Molecular Weight (g/mol):
220.15
Molecular Formula:
C8H7F3N2O2
InChi Key:
KGHGHEPVWUOJAZ-UHFFFAOYSA-N
SMILES:
NC(=NO)C1=CC=CC=C1OC(F)(F)F
1
–
1
of
1
results
1
N'-Hydroxy-2-(trifluoromethoxy)benzenecarboximidamide, 95%, Thermo Scientific™
CAS: 63968-84-3 Molecular Formula: C8H7F3N2O2 Molecular Weight (g/mol): 220.15 MDL Number: MFCD07772878 InChI Key: KGHGHEPVWUOJAZ-UHFFFAOYSA-N Synonym: 2-trifluoromethoxy benzamidoxime,ho dpmlgbprprprjpjjiqy jhhfhabp,n'-hydroxy-2-trifluoromethoxy benzimidamide,hydroxyimino 2-trifluoromethoxy phenyl methylamine,benzenecarboximidamide, n-hydroxy-2-trifluoromethoxy,z-n'-hydroxy-2-trifluoromethoxy benzene-1-carboximidamide PubChem CID: 9582829 SMILES: NC(=NO)C1=CC=CC=C1OC(F)(F)F