missing translation for 'onlineSavingsMsg'
Learn More

N'-Hydroxy-2-(trifluoromethoxy)benzenecarboximidamide, 95%, Thermo Scientific™

Product Code. 10167232
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10167232 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10167232 Supplier Thermo Scientific Maybridge Supplier No. MO01204EA
 Restricted Item
Estimated Shipment: 11-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 63968-84-3
Molecular Formula C8H7F3N2O2
Molecular Weight (g/mol) 220.15
MDL Number MFCD07772878
InChI Key KGHGHEPVWUOJAZ-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy benzamidoxime, ho dpmlgbprprprjpjjiqy jhhfhabp, n'-hydroxy-2-trifluoromethoxy benzimidamide, hydroxyimino 2-trifluoromethoxy phenyl methylamine, benzenecarboximidamide, n-hydroxy-2-trifluoromethoxy, z-n'-hydroxy-2-trifluoromethoxy benzene-1-carboximidamide
PubChem CID 9582829
SMILES NC(=NO)C1=CC=CC=C1OC(F)(F)F

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
IUPAC Name N'-hydroxy-2-(trifluoromethoxy)benzenecarboximidamide
Formula Weight 220.15
Percent Purity 95%
Chemical Name or Material N'-hydroxy-2-(trifluoromethoxy)benzenecarboximidamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.