accessibility menu, dialog, popup

UserName

FFN

CAS RN 64248-63-1

IUPAC Name: 3,5-difluorobenzonitrile
Synonyms: 3,5-difluorobenzonitrile
Molecular Weight (g/mol): 139.11
Molecular Formula: C7H3F2N
InChi Key: CQXZSEXZQVKCHW-UHFFFAOYSA-N
SMILES: FC1=CC(=CC(F)=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results