accessibility menu, dialog, popup

UserName

H3COHOOO

CAS RN 645-08-9

IUPAC Name: 3-hydroxy-4-methoxybenzoate
Synonyms: isovanillate
Molecular Weight (g/mol): 167.14
Molecular Formula: C8H7O4
InChi Key: LBKFGYZQBSGRHY-UHFFFAOYSA-M
SMILES: COC1=CC=C(C=C1O)C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results