accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 646-31-1

IUPAC Name: tetracosane
Synonyms: tetracosane
Molecular Weight (g/mol): 338.66
Molecular Formula: C24H50
InChi Key: POOSGDOYLQNASK-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (2)
  • (4)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (4)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)