accessibility menu, dialog, popup

UserName

P

CAS RN 6476-37-5

IUPAC Name: dicyclohexyl(phenyl)phosphane
Synonyms: dicyclohexylphenylphosphine
Molecular Weight (g/mol): 274.39
Molecular Formula: C18H27P
InChi Key: VPLLTGLLUHLIHA-UHFFFAOYSA-N
SMILES: C1CCC(CC1)P(C1CCCCC1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results