missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexylphenylphosphine, 95%, Thermo Scientific Chemicals

Product Code. 10528892 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10528892 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10528892 Supplier Thermo Scientific Acros Supplier No. 407080010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6476-37-5
Molecular Formula C18H27P
Molecular Weight (g/mol) 274.39
MDL Number MFCD00003854
InChI Key VPLLTGLLUHLIHA-UHFFFAOYSA-N
Synonym dicyclohexylphenylphosphine, dicyclohexyl phenyl phosphine, phosphine, dicyclohexylphenyl, phenyldicyclohexylphosphine, phenylphosphinodicyclohexane, dicyclohexyl phenyl phosphane, pphcy2, acmc-1bey7, dicyclohexyl-phenyl-phosphane
PubChem CID 80970
IUPAC Name dicyclohexyl(phenyl)phosphane
SMILES C1CCC(CC1)P(C1CCCCC1)C1=CC=CC=C1

Specifications

Melting Point 54.0°C to 57.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 1 g
Formula Weight 274.38
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Dicyclohexylphenylphosphine
compliance-icons
Product Identifier
  • Dicyclohexylphenylphosphine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.