accessibility menu, dialog, popup

UserName

OO

CAS RN 6485-97-8

IUPAC Name: 2-phenyl-9,10-dihydroanthracene-9,10-dione
Synonyms: 2-phenylanthracene-9,10-dione
Molecular Weight (g/mol): 284.31
Molecular Formula: C20H12O2
InChi Key: NTZCFGZBDDCNHI-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2C(=O)C2=CC(=CC=C12)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results