accessibility menu, dialog, popup

UserName

H3CS

CAS RN 65016-62-8

IUPAC Name: 3-octylthiophene
Synonyms: 3-octylthiophene
Molecular Weight (g/mol): 196.35
Molecular Formula: C12H20S
InChi Key: WQYWXQCOYRZFAV-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CSC=C1
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)