missing translation for 'onlineSavingsMsg'
Learn More

3-n-Octylthiophene, 97%

Product Code. 15491718
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491718 5 g 5g
15481718 1 g 1g
15401728 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491718 Supplier Thermo Scientific Alfa Aesar Supplier No. H55466.06
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 65016-62-8 | C12H20S | 196.352 g/mol

3-Octylthiophene is used as a starting material in the synthesis of the following 2,5-dibromo-3-octylthiophene, poly(3-butylthiophene)-b-poly(3-octylthiophene), a diblock copoly(3-alkylthiophene), regioregular poly(3-octylthiophene).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Octylthiophene is used as a starting material in the synthesis of the following 2,5-dibromo-3-octylthiophene, poly(3-butylthiophene)-b-poly(3-octylthiophene), a diblock copoly(3-alkylthiophene), regioregular poly(3-octylthiophene).

Solubility
Soluble in organic solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65016-62-8
Molecular Formula C12H20S
Molecular Weight (g/mol) 196.352
MDL Number MFCD00085281
InChI Key WQYWXQCOYRZFAV-UHFFFAOYSA-N
Synonym 3-n-octylthiophene, thiophene, 3-octyl, acmc-1bfcd, 3-octylthiophene, ksc491i4b, bidd:gt0691
PubChem CID 566852
IUPAC Name 3-octylthiophene
SMILES CCCCCCCCC1=CSC=C1

Specifications

Color Colorless
Density 0.92 g/mL
Boiling Point 106°C to 107°C (3 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.492
Quantity 5 g
Solubility Information Soluble in organic solvents.
Formula Weight 196.35
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-n-Octylthiophene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.