accessibility menu, dialog, popup

UserName

CAS RN 651-80-9

FFFFFFF

CAS RN 651-80-9

IUPAC Name: 1,2,4,5-tetrafluoro-3-(trifluoromethyl)benzene
Synonyms: 1,2,4,5-tetrafluoro-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 218.07
Molecular Formula: C7HF7
InChi Key: ZVPAJILXQHMKMT-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(F)C(=C1F)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results