accessibility menu, dialog, popup

UserName

HOOO

CAS RN 6515-36-2

IUPAC Name: 7-hydroxy-2-phenyl-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: 7-hydroxyflavanone
Molecular Weight (g/mol): 240.26
Molecular Formula: C15H12O3
InChi Key: SWAJPHCXKPCPQZ-UHFFFAOYNA-N
SMILES: OC1=CC=C2C(=O)CC(OC2=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)