accessibility menu, dialog, popup

UserName

CAS RN 6515-37-3

HOOO

CAS RN 6515-37-3

IUPAC Name: 2-(4-hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: 4'-hydroxyflavanone
Molecular Weight (g/mol): 240.26
Molecular Formula: C15H12O3
InChi Key: ZLHVIYHWWQYJID-UHFFFAOYNA-N
SMILES: OC1=CC=C(C=C1)C1CC(=O)C2=CC=CC=C2O1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)