CAS RN 65201-77-6
CAS RN 65201-77-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tetrabutylammonium Periodate, TRC
CAS: 65201-77-6 Molecular Formula: C16H36N . IO4 Molecular Weight (g/mol): 433.37 Synonym: N,N,N-Tributyl-1-butanaminium Periodate,N,N,N-Tributyl-1-butanaminium Salt With Periodic Acid (HIO4),N,N,N-Tributyl-1-butanaminium Periodate (IO41-),Tetrabutylammonium Metaperiodate,Tetrabutylammonium Periodate IUPAC Name: tetrabutylazanium;periodate SMILES: CCCC[N+](CCCC)(CCCC)CCCC.[O-]I(=O)(=O)=O