missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Periodate, TRC

Product Code. 30414157
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30414157 10 g 10g
30411941 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30414157 Supplier Toronto Research Chemicals Supplier No. TRCT28923010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65201-77-6
Molecular Formula C16H36N . IO4
Molecular Weight (g/mol) 433.37
Synonym N,N,N-Tributyl-1-butanaminium Periodate, N,N,N-Tributyl-1-butanaminium Salt With Periodic Acid (HIO4), N,N,N-Tributyl-1-butanaminium Periodate (IO41-), Tetrabutylammonium Metaperiodate, Tetrabutylammonium Periodate
IUPAC Name tetrabutylazanium;periodate
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[O-]I(=O)(=O)=O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C16H36N.HIO4/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;2-1(3,4)5/h5-16H2,1-4H3;(H,2,3,4,5)/q+1;/p-1
Formula Weight 433.1689
Chemical Name or Material Tetrabutylammonium Periodate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.