accessibility menu, dialog, popup

UserName

NH2ONHF

CAS RN 656-31-5

IUPAC Name: (2-fluorophenyl)urea
Synonyms: O-fluorophenylurea
Molecular Weight (g/mol): 154.14
Molecular Formula: C7H7FN2O
InChi Key: PAWVOCWEWJXILY-UHFFFAOYSA-N
SMILES: NC(=O)NC1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results