accessibility menu, dialog, popup

UserName

OOOO

CAS RN 65709-23-1

IUPAC Name: 2-(2H-1,3-benzodioxol-5-yl)-2-oxoacetaldehyde hydrate
Synonyms: 2-(2H-1,3-benzodioxol-5-yl)-2-oxoacetaldehyde hydrate
Molecular Weight (g/mol): 196.16
Molecular Formula: C9H8O5
InChi Key: ZETZLJWJCYEROP-UHFFFAOYSA-N
SMILES: O.O=CC(=O)C1=CC2=C(OCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results