missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)phenylglyoxal hydrate, 97%, dry wt basis

Product Code. 11389136
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11389136 5 g 5g
11379136 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389136 Supplier Thermo Scientific Alfa Aesar Supplier No. L20273.06
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 65709-23-1 | C9H6O4 | 178.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65709-23-1
Molecular Formula C9H6O4
Molecular Weight (g/mol) 178.14
MDL Number MFCD03411518
InChI Key OUCBKIWZWGSDJL-UHFFFAOYSA-N
Synonym 3,4-methylenedioxyphenylglyoxal hydrate, 2-benzo d 1,3 dioxol-5-yl-2-oxoacetaldehyde hydrate, 3,4-methylenedioxy phenylglyoxal hydrate, 2-2h-1,3-benzodioxol-5-yl-2-oxoacetaldehyde hydrate, pubchem12143, benzo d 1,3 dioxole-5-glyoxal hydrate, 2-1,3-benzodioxol-5-yl-2-oxoacetaldehyde hydrate, 2-1,3-benzodioxol-5-yl-2-oxidanylidene-ethanal hydrate, 2-2h-benzo 3,4-d 1,3-dioxolan-5-yl-2-oxoethanal, hydrate
PubChem CID 15608490
IUPAC Name 2-(1,3-benzodioxol-5-yl)-2-oxoacetaldehyde;hydrate
SMILES O=CC(=O)C1=CC2=C(OCO2)C=C1

Specifications

Melting Point 127°C to 131°C
Quantity 5 g
Beilstein 7703971
Formula Weight 178.15 (Anhydrous)
Percent Purity 97%
Assay (dry wt.)
Chemical Name or Material 3,4-(Methylenedioxy)phenylglyoxal hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.