accessibility menu, dialog, popup

UserName

CAS RN 65796-00-1

ClOFFF

CAS RN 65796-00-1

IUPAC Name: 1-(chloromethyl)-4-(trifluoromethoxy)benzene
Synonyms: 1-(chloromethyl)-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 210.58
Molecular Formula: C8H6ClF3O
InChi Key: LBMKFQMJURUPKC-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(CCl)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results