accessibility menu, dialog, popup

UserName

CAS RN 66191-86-4

H3COOBr

CAS RN 66191-86-4

IUPAC Name: methyl 3-(2-bromophenyl)propanoate
Synonyms: methyl 3-(2-bromophenyl)propanoate
Molecular Weight (g/mol): 243.1
Molecular Formula: C10H11BrO2
InChi Key: NIULOVGPRAJIJM-UHFFFAOYSA-N
SMILES: COC(=O)CCC1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results