accessibility menu, dialog, popup

UserName

OHBrHO

CAS RN 6626-15-9

IUPAC Name: 4-bromobenzene-1,3-diol
Synonyms: 4-bromoresorcinol
Molecular Weight (g/mol): 189.01
Molecular Formula: C6H5BrO2
InChi Key: MPCCNXGZCOXPMG-UHFFFAOYSA-N
SMILES: OC1=CC=C(Br)C(O)=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)