accessibility menu, dialog, popup

UserName

CAS RN 6630-65-5

Cl

CAS RN 6630-65-5

IUPAC Name: 9-chloro-9H-fluorene
Synonyms: 9-chloro-9H-fluorene
Molecular Weight (g/mol): 200.67
Molecular Formula: C13H9Cl
InChi Key: CVCQMAKDIKSUHX-UHFFFAOYSA-N
SMILES: ClC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)