accessibility menu, dialog, popup

UserName

NH2Br

CAS RN 6638-60-4

IUPAC Name: 7-bromo-9H-fluoren-2-amine
Synonyms: 9H-fluoren-2-amine, 7-bromo-
Molecular Weight (g/mol): 260.13
Molecular Formula: C13H10BrN
InChi Key: RJWYTISHBMNMOZ-UHFFFAOYSA-N
SMILES: NC1=CC=C2C(CC3=CC(Br)=CC=C23)=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)