accessibility menu, dialog, popup

UserName

H3CH3CCH3O

CAS RN 6669-13-2

IUPAC Name: (tert-butoxy)benzene
Synonyms: benzene, (1,1-dimethylethoxy)-
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: PNKZBZPLRKCVLI-UHFFFAOYSA-N
SMILES: CC(C)(C)OC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results