accessibility menu, dialog, popup

UserName

CAS RN 66778-08-3

IUPAC Name: phenoxyphosphonic acid dihydrate disodium
Synonyms: dihydrate phenylphosphate disodium
Molecular Weight (g/mol): 256.1
Molecular Formula: C6H11Na2O6P
InChi Key: UMRLIPYPVZYYFT-UHFFFAOYSA-N
SMILES: O.O.[Na].[Na].OP(O)(=O)OC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results