missing translation for 'onlineSavingsMsg'
Learn More

Phenyl phosphate, disodium salt dihydrate, 98%

Product Code. 10318350 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10318350 100 g Glass bottle 100g
10184690 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10318350 Supplier Thermo Scientific Acros Supplier No. 211831000
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phenyl phosphate, disodium salt dihydrate, 98%, C6H5Na2O4P.2H2O, CAS Number-66778-08-3, sodiumphenylphosphatedibasicdihydrate, 10g, White, 221-917-4, 254.09, HYGROSCOPIC, UMRLIPYPVZYYFT-UHFFFAOYSA-N, Authentic, phenyl dihydrogen phosphate;sodium;dihydrate, C6H5OP(O)(ONa)2.2H2O, >300.0 deg.C | CAS: 66778-08-3 | C6H5Na2O42H2O | 254.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66778-08-3
Molecular Formula C6H5Na2O42H2O
Molecular Weight (g/mol) 254.09
MDL Number MFCD00149080
InChI Key UMRLIPYPVZYYFT-UHFFFAOYSA-N
Synonym sodiumphenylphosphatedibasicdihydrate
PubChem CID 131883070
IUPAC Name phenyl dihydrogen phosphate;sodium;dihydrate
SMILES C1=CC=C(C=C1)OP(=O)(O)O.O.O.[Na].[Na]

Specifications

Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5OP(O)(ONa)2·2H2O
Quantity 100 g
Merck Index 14, 3357
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and chloroform,insoluble in acetone and ether
Formula Weight 254.09
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Phenyl phosphate, disodium salt dihydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.