accessibility menu, dialog, popup

UserName

CAS RN 670-96-2

IUPAC Name: 2-phenyl-1H-imidazole
Synonyms: 1H-imidazole, 2-phenyl-
Molecular Weight (g/mol): 144.18
Molecular Formula: C9H8N2
InChi Key: ZCUJYXPAKHMBAZ-UHFFFAOYSA-N
SMILES: N1C=CN=C1C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)