missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylimidazole, 98%

Product Code. 11426706
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426706 25 g 25g
11436706 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11426706 Supplier Thermo Scientific Alfa Aesar Supplier No. B23858.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 670-96-2 | C9H8N2 | 144.18 g/mol

2-Phenylimidazole is used to prepare complex compounds with ruthenium(III) and also as an intermediate for agrochemical, pharmaceutical and specialty products. It also finds its application as a curing agent for epoxy resins and polyurethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenylimidazole is used to prepare complex compounds with ruthenium(III) and also as an intermediate for agrochemical, pharmaceutical and specialty products. It also finds its application as a curing agent for epoxy resins and polyurethane.

Solubility
Solubility in methanol (almost transparency).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 670-96-2
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00005186
InChI Key ZCUJYXPAKHMBAZ-UHFFFAOYSA-N
Synonym 2-phenylimidazole, 1h-imidazole, 2-phenyl, imidazole, 2-phenyl, 2-phenyl imidazole, 2-phenyl-1h-imidazol, unii-7r2590hm4l, chembl14189, phenyl imidazole, 2-phenyl-imidazole, piy
PubChem CID 69591
IUPAC Name 2-phenyl-1H-imidazole
SMILES N1C=CN=C1C1=CC=CC=C1

Specifications

Melting Point 146°C to 150°C
Boiling Point 198°C to 200°C (7 mmHg)
Flash Point 200°C (392°F)
Quantity 25 g
Beilstein 114011
Formula Weight 144.18
Percent Purity ≥98%
Chemical Name or Material 2-Phenylimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.