accessibility menu, dialog, popup

UserName

H3COOOH3C

CAS RN 672-89-9

IUPAC Name: 4-methoxy-6-methyl-2H-pyran-2-one
Synonyms: 4-methoxy-6-methylpyran-2-one
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: MTZAUZNQAMNFME-UHFFFAOYSA-N
SMILES: COC1=CC(=O)OC(C)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)