missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-6-methyl-2H-pyran-2-one, 97%, Thermo Scientific Chemicals

Product Code. 15417928
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15417928 1 g 1g
15427928 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417928 Supplier Thermo Scientific Alfa Aesar Supplier No. H61395.03
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 672-89-9 | Molecular Formula: C7H8O3 | Molecular Weight: 140.14 g/mol

4-Methoxy-6-methyl-2H-pyran-2-one is used as a model compound to study the assignment of ring proton resonances in 4-methoxy-6-nonyl-2H-pyran-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-6-methyl-2H-pyran-2-one is used as a model compound to study the assignment of ring proton resonances in 4-methoxy-6-nonyl-2H-pyran-2-one.

Solubility
Poorly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 672-89-9
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.14
MDL Number MFCD00006640
InChI Key MTZAUZNQAMNFME-UHFFFAOYSA-N
Synonym 4-methoxy-6-methyl-2h-pyran-2-one, methyltriacetolactone, methyltriacetolacton, 2h-pyran-2-one, 4-methoxy-6-methyl, 4-methoxy-6-methyl-pyran-2-one, methyltriacetolacton german, 4-methoxy-6-methyl-2-pyrone, 4mhp, wln: t6ovj do1 f1, 4-methoxy-6-methyl-2-pyranone
PubChem CID 12651
IUPAC Name 4-methoxy-6-methylpyran-2-one
SMILES COC1=CC(=O)OC(C)=C1

Specifications

Melting Point 83°C to 86°C
Quantity 1 g
Solubility Information Poorly soluble in water.
Formula Weight 140.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Methoxy-6-methyl-2H-pyran-2-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.