CAS RN 67247-13-6
CAS RN 67247-13-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7-Methoxy-1-naphthalenol, TRC
CAS: 67247-13-6 Molecular Formula: C11 H10 O2 Molecular Weight (g/mol): 174.2 IUPAC Name: 7-methoxynaphthalen-1-ol SMILES: COc1ccc2cccc(O)c2c1