CAS RN 67375-30-8
IUPAC Name:
cyano(3-phenoxyphenyl)methyl (1R,3R)-3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
Synonyms:
cyano(3-phenoxyphenyl)methyl (1R,3R)-3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
Molecular Weight (g/mol):
416.3
Molecular Formula:
C22H19Cl2NO3
InChi Key:
KAATUXNTWXVJKI-DXCJPMOASA-N
SMILES:
CC1(C)[C@@H](C=C(Cl)Cl)[C@H]1C(=O)OC(C#N)C1=CC=CC(OC2=CC=CC=C2)=C1
1
–
8
of
8
results
2
alpha-Cypermethrin (Racemic Mixture), TRC
CAS: 67375-30-8 Molecular Formula: C22 H19 Cl2 N O3 Molecular Weight (g/mol): 416.3 Synonym: (±)-Alphamethrin,Aeralfam,Alfazot,Alfoxylate,Alphacypermethrin,Alphaguard,Alphamethrin,Astound Duo,BAS 310,Bestox,Centaur,Cismix,Contest,Dominex,Dudu Alpha,Fastac,Fastac 100EC,Fastac 10EC,Fastac CS,Fastac Duo,Fendona,Fendona 60SC,Fendona 65C,Mageos,Sentry,Sentry (insecticide),Ultimate,WL 85871,α-Cypermethrin IUPAC Name: [(R)-cyano-(3-phenoxyphenyl)methyl] (1S,3S)-3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate SMILES: CC1(C)[C@H](C=C(Cl)Cl)[C@@H]1C(=O)O[C@@H](C#N)c2cccc(Oc3ccccc3)c2