CAS RN 67376-94-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Methylcyclopropanamine Hydrochloride, TRC
CAS: 67376-94-7 Molecular Formula: C4 H9 N . H Cl Molecular Weight (g/mol): 107.58 Synonym: (Methyl)(Cyclopropyl)Amine Hydrochloride,N-Cyclopropylmethylamine Hydrochloride IUPAC Name: N-methylcyclopropanamine;hydrochloride SMILES: Cl.Cnc1Cc1