missing translation for 'onlineSavingsMsg'
Learn More

N-Methylcyclopropanamine Hydrochloride, TRC

Product Code. 30482721
Change view
Click to view available options
Quantity:
250 mg
2.5 g
5 g
Unit Size:
2.50g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482721 2.5 g 2.50g
30465930 5 g 5g
30481688 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482721 Supplier Toronto Research Chemicals Supplier No. TRCM2950432.5G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67376-94-7
Molecular Formula C4 H9 N . H Cl
Molecular Weight (g/mol) 107.58
Synonym (Methyl)(Cyclopropyl)Amine Hydrochloride, N-Cyclopropylmethylamine Hydrochloride
IUPAC Name N-methylcyclopropanamine;hydrochloride
SMILES Cl.Cnc1Cc1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C4H9N.ClH/c1-5-4-2-3-4;/h4-5H,2-3H2,1H3;1H
Formula Weight 107.0502
Chemical Name or Material N-Methylcyclopropanamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.