accessibility menu, dialog, popup

UserName

CAS RN 67531-86-6

FOOHHO

CAS RN 67531-86-6

IUPAC Name: 2-fluoro-6-hydroxybenzoic acid
Synonyms: 2-fluoro-6-hydroxybenzoic acid
Molecular Weight (g/mol): 156.11
Molecular Formula: C7H5FO3
InChi Key: BCEKGWWLVKXZKK-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(O)C=CC=C1F
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results