CAS RN 675602-89-8
IUPAC Name:
3,6-difluoro-4-(trifluoromethyl)pyridin-2-amine
Synonyms:
3,6-difluoro-4-(trifluoromethyl)pyridin-2-amine
Molecular Weight (g/mol):
198.1
Molecular Formula:
C6H3F5N2
InChi Key:
GJYDRBORZUFFEB-UHFFFAOYSA-N
SMILES:
NC1=C(F)C(=CC(F)=N1)C(F)(F)F
1
–
1
of
1
results
1
2-Amino-3,6-difluoro-4-(trifluoromethyl)pyridine, 95%, Thermo Scientific™
CAS: 675602-89-8 Molecular Formula: C6H3F5N2 Molecular Weight (g/mol): 198.096 MDL Number: MFCD03840200 InChI Key: GJYDRBORZUFFEB-UHFFFAOYSA-N Synonym: 2-amino-3,6-difluoro-4-trifluoromethyl pyridine,3,6-difluoro-4-trifluoromethyl pyridin-2-amine,2-amino-3,6-difluoro-4-trifluoromethyl-pyridine,3,6-difluoro-4-trifluoromethyl-2-pyridylamine,pubchem2973,3,6-difluoro-4-trifluoromethyl-2-pyridinamine,2-pyridinamine,3,6-difluoro-4-trifluoromethyl,3,6-bis fluoranyl-4-trifluoromethyl pyridin-2-amine PubChem CID: 2782809 IUPAC Name: 3,6-difluoro-4-(trifluoromethyl)pyridin-2-amine SMILES: C1=C(C(=C(N=C1F)N)F)C(F)(F)F