missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,6-difluoro-4-(trifluoromethyl)pyridine, 95%, Thermo Scientific™

Product Code. 11374396
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374396 100 mg 100mg
11364396 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374396 Supplier Thermo Scientific Alfa Aesar Supplier No. L20009.MC
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 675602-89-8
Molecular Formula C6H3F5N2
Molecular Weight (g/mol) 198.096
MDL Number MFCD03840200
InChI Key GJYDRBORZUFFEB-UHFFFAOYSA-N
Synonym 2-amino-3,6-difluoro-4-trifluoromethyl pyridine, 3,6-difluoro-4-trifluoromethyl pyridin-2-amine, 2-amino-3,6-difluoro-4-trifluoromethyl-pyridine, 3,6-difluoro-4-trifluoromethyl-2-pyridylamine, pubchem2973, 3,6-difluoro-4-trifluoromethyl-2-pyridinamine, 2-pyridinamine,3,6-difluoro-4-trifluoromethyl, 3,6-bis fluoranyl-4-trifluoromethyl pyridin-2-amine
PubChem CID 2782809
IUPAC Name 3,6-difluoro-4-(trifluoromethyl)pyridin-2-amine
SMILES C1=C(C(=C(N=C1F)N)F)C(F)(F)F

Specifications

Melting Point 66°C to 68°C
Quantity 100 mg
UN Number UN2811
Formula Weight 198.1
Percent Purity 95%
Chemical Name or Material 2-Amino-3,6-difluoro-4-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.