accessibility menu, dialog, popup

UserName

CAS RN 67589-38-2

CH3OOOCH3

CAS RN 67589-38-2

IUPAC Name: 4-methoxyphenyl 4-propylcyclohexane-1-carboxylate
Synonyms: 4-methoxyphenyl 4-propylcyclohexane-1-carboxylate
Molecular Weight (g/mol): 276.38
Molecular Formula: C17H24O3
InChi Key: YOMOGIXKVWWEHG-UHFFFAOYSA-N
SMILES: CCCC1CCC(CC1)C(=O)OC1=CC=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results