accessibility menu, dialog, popup

UserName

H3COOCH3NHSNH2

CAS RN 67617-98-5

IUPAC Name: (2,5-dimethoxyphenyl)thiourea
Synonyms: 2,5-dimethoxyphenylthiourea
Molecular Weight (g/mol): 212.27
Molecular Formula: C9H12N2O2S
InChi Key: ZLGQKHQZDDTIFW-UHFFFAOYSA-N
SMILES: COC1=CC(NC(N)=S)=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results