accessibility menu, dialog, popup

UserName

H3CMgClCH3CH3

CAS RN 677-22-5

IUPAC Name: magnesium(2+) 2-methylpropan-2-ide chloride
Synonyms: magnesium(2+) 2-methylpropan-2-ide chloride
Molecular Weight (g/mol): 116.87
Molecular Formula: C4H9ClMg
InChi Key: CQRPUKWAZPZXTO-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].C[C-](C)C
Quantity 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)