accessibility menu, dialog, popup

UserName

FFFFIFFF

CAS RN 677-69-0

IUPAC Name: 1,1,1,2,3,3,3-heptafluoro-2-iodopropane
Synonyms: 1,1,1,2,3,3,3-heptafluoro-2-iodopropane
Molecular Weight (g/mol): 295.93
Molecular Formula: C3F7I
InChi Key: BBZVTTKMXRPMHZ-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(I)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)